Learn More
Zeiss™ Ocular Micrometer
Product Code. 11868422
Change view
Click to view available options
Diameter (Metric):
23 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
11868422 23 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11868422 Supplier Zeiss™ Supplier No. 4155001813000
This item requires a price update prior to being ordered.
Request a Price for this item here.
Designed for use with Zeiss Microscopes
  • Crossline micrometer
  • 10:100
  • diam: 23mm
  • For Primo Microscopes
Specifications
Type Micrometer
Diameter (Metric) 23 mm
For Use With (Equipment) Primo Microscopes
Quantity 1 Ea.