missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ODF2L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23887-1-AP-20UL
This item is not returnable.
View return policy
Description
ODF2L, also named as KIAA1229, has many isoforms with MW 60-80 kDa. The function of ODF2L (Outer dense fiber protein 2-like) remains largely unknown.Specifications
| ODF2L | |
| Polyclonal | |
| Unconjugated | |
| ODF2L | |
| KIAA1229, ODF2L, RP5 977L11.1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 52184, 57489, 685425 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9D478, Q9ULJ1 | |
| Odf2l | |
| ODF2L Fusion Protein Ag20897 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction