Learn More
OFD1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 22851-1-AP-20UL
 View more versions of this product
Product Code. 16802115
  • 145.00€ / 20µL
Description
This antibody recognize all the isoforms of OFD1. The 43 kDa is isoform 2. The ∽80 kDa band is unknown.
This gene is located on the X chromosome and encodes a centrosomal protein. A knockout mouse model has been used to study the effect of mutations in this gene. The mouse gene is also located on the X chromosome, however, unlike the human gene it is not subject to X inactivation. Mutations in this gene are associated with oral-facial-digital syndrome type I and Simpson-Golabi-Behmel syndrome type 2. Many pseudogenes have been identified; a single pseudogene is found on chromosome 5 while as many as fifteen have been found on the Y chromosome.
Specifications
Specifications
OFD1
Polyclonal
Unconjugated
OFD1
71 7A, CXorf5, OFD1, oral facial digital syndrome 1, Protein 71 7A, SGBS2
Rabbit
Antigen Affinity Chromatography
RUO
8481
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.29 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O75665
OFD1
OFD1 Fusion Protein Ag18898