missing translation for 'onlineSavingsMsg'
Learn More

Ofloxacin, Thermo Scientific™

Fluorinated nalidixic acid analog with broad-spectrum antibacterial activity

Brand:  Thermo Scientific Alfa Aesar J62080.03

Quantity 1g

 View more versions of this product

Product Code. 15454529

  • ONLINE EXCLUSIVE PRICE
    39.40€
    38.59€ / 1g
    Save 0.81€
    2% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

82419-36-1
361.373
GSDSWSVVBLHKDQ-UHFFFAOYSA-N
4583
CC1COC2=C3N1C=C(C(=O)C3=CC(=C2N4CCN(CC4)C)F)C(=O)O
C18H20FN3O4
MFCD00226105
Oflocet; Ofloxacine
CHEBI:7731

Specifications

82419-36-1
1g
MFCD00226105
Oflocet; Ofloxacine
GSDSWSVVBLHKDQ-UHFFFAOYSA-N
361.373
CHEBI:7731
Ofloxacin
White to Yellow
C18H20FN3O4
14,6771
Soluble in acetic acid or water; Slightly soluble in methanol
CC1COC2=C3N1C=C(C(=O)C3=CC(=C2N4CCN(CC4)C)F)C(=O)O
4583
361.37
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.