Learn More
Ofloxacin, Thermo Scientific™
Fluorinated nalidixic acid analog with broad-spectrum antibacterial activity
38.59€ - 138.00€
Chemical Identifiers
| CAS | 82419-36-1 |
|---|---|
| Molecular Formula | C18H20FN3O4 |
| Molecular Weight (g/mol) | 361.373 |
| MDL Number | MFCD00226105 |
| InChI Key | GSDSWSVVBLHKDQ-UHFFFAOYSA-N |
| Synonym | Oflocet; Ofloxacine |
| PubChem CID | 4583 |
| ChEBI | CHEBI:7731 |
| SMILES | CC1COC2=C3N1C=C(C(=O)C3=CC(=C2N4CCN(CC4)C)F)C(=O)O |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15454529
|
Thermo Scientific Alfa Aesar
J62080.03 |
1g |
39.40€ 38.59€ / 1g Save 0.81€ 2% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15464529
|
Thermo Scientific Alfa Aesar
J62080.06 |
5g |
141.00€ 138.00€ / 5g Save 3.00€ 2% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 82419-36-1 | |
| 361.373 | |
| GSDSWSVVBLHKDQ-UHFFFAOYSA-N | |
| 4583 | |
| CC1COC2=C3N1C=C(C(=O)C3=CC(=C2N4CCN(CC4)C)F)C(=O)O |
| C18H20FN3O4 | |
| MFCD00226105 | |
| Oflocet; Ofloxacine | |
| CHEBI:7731 |
Specifications
| 82419-36-1 | |
| C18H20FN3O4 | |
| 14,6771 | |
| Soluble in acetic acid or water; Slightly soluble in methanol | |
| CC1COC2=C3N1C=C(C(=O)C3=CC(=C2N4CCN(CC4)C)F)C(=O)O | |
| 4583 | |
| 361.37 |
| White to Yellow | |
| MFCD00226105 | |
| Oflocet; Ofloxacine | |
| GSDSWSVVBLHKDQ-UHFFFAOYSA-N | |
| 361.373 | |
| CHEBI:7731 | |
| Ofloxacin |
Safety and Handling
missing translation for 'rtecsNumber' : UU8815500
missing translation for 'tsca' : No
Recommended Storage : Store at -20°C
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.