missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Oil Red O

Product Code. p-7020882
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11459030 25 g 25g
11469030 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11459030 Supplier Thermo Scientific Chemicals Supplier No. A12989.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1320-06-5 | C26H24N4O | 408.51 g/mol

Oil Red O is suitable as a lipid and lipoprotein stain on cellulose acetate. It is one of the diazo dyes used for staining of neutral triglycerides and lipids. It is used to analyze muscle fibers. It acts as an agent that stains lipids, muscle fibers and alveolar macrophages. Further, it is also used as marking dye. It is an active component of red colored smokes in pyrotechnics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Oil Red O is suitable as a lipid and lipoprotein stain on cellulose acetate. It is one of the diazo dyes used for staining of neutral triglycerides and lipids. It is used to analyze muscle fibers. It acts as an agent that stains lipids, muscle fibers and alveolar macrophages. Further, it is also used as marking dye. It is an active component of red colored smokes in pyrotechnics.

Solubility
Soluble in ethanol:chloroform (1:1). Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1320-06-5
Molecular Formula C26H24N4O
Molecular Weight (g/mol) 408.51
MDL Number MFCD00003898
InChI Key HJAYEODIXUYVIC-LTFAAIOASA-N
Synonym C.I. 26125; Solvent Red 27
PubChem CID 6046885
IUPAC Name (1Z)-1-[[4-[(2,5-dimethylphenyl)diazenyl]-2,5-dimethylphenyl]hydrazinylidene]naphthalen-2-one
SMILES CC1=CC=C(C)C(=C1)N=NC1=CC(C)=C(N\N=C2/C(=O)C=CC3=CC=CC=C23)C=C1C

Specifications

Melting Point ∼166°C (decomposition)
Beilstein 3491382
Solubility Information Soluble in ethanol:chloroform (1:1). Slightly soluble in water.
Formula Weight 408.51
Quantity 25 g
Chemical Name or Material Oil Red O

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.