Description
- Fast and easy to use - No training or special equipment required
- Supports HACCP and food safety compliance
- Improves food presentation and taste
- Reduces oil waste and fryer maintenance costs
- Trusted by chefs and kitchen managers across the UK and Europe
- Easy-to-Use: Designed with simplicity in mind, these test strips offer a hassle-free testing experience. With a user-friendly dip-and-read method, chefs and home cooks can quickly assess the quality of their cooking oil without the need for specialized equipment or extensive training.
- High Sensitivity: Engineered to detect various indicators of cooking oil degradation, these test strips offer unparalleled sensitivity. From Free Fatty Acid (FFA) levels to Total Polar Compounds (TPC), users can obtain comprehensive insights into the quality and freshness of their cooking oil.
- Rapid Results: With rapid response times, users can obtain test results within 30 seconds, allowing for timely intervention and proactive oil management. Whether it's determining the optimal time for oil replacement or monitoring oil stability during frying, these test strips empower users to maintain peak oil performance.
- Versatility: Suitable for a wide range of cooking oils, including vegetable oil, olive oil, canola oil, and more, these testing strips offer versatility and adaptability to diverse culinary applications.
- Cost-Effective: By enabling precise oil management and extending the lifespan of cooking oil, these test strips help reduce waste and optimize operational costs for food service establishments. Their affordability and efficiency make them a valuable investment for kitchens of all sizes.
Specifications
Specifications
| Detects | Oil |
| Manufacturing Origin | United Kingdom |
| Product Type | Oil Test Strip |
| Test Type | Qualitative QC |
| Quantity | 1 Ea. |
Product Title
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?