missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oleic acid-glycidyl ester 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Glycidyl Oleate |
| CAS | 5431-33-4 |
| Analyte or Component Names | Glycidyl Oleate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H38 O3 |
| Concentration | 100 μg/mL |
| Certifications/Compliance | ISO 17034 |
| Synonym | (9Z)-9-Octadecenoic Acid 2-Oxiranylmethyl Ester, Oleic Acid 2,3-Epoxypropyl Ester, NSC 13542, Oleic Acid Glycidyl Ester; |
| InChI Formula | InChI=1S/C21H38O3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-21(22)24-19-20-18-23-20/h9-10,20H,2-8,11-19H2,1H3/b10-9- |
| SMILES | CCCCCCCC\C=C/CCCCCCCC(=O)OCC1CO1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?