missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OLFM3 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
180.00€ - 453.00€
Specifications
| Antigen | OLFM3 |
|---|---|
| Concentration | 0.5 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17753124
|
Invitrogen™
PA5121727 |
20 μL |
180.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17743124
![]() |
Invitrogen™
PA5121726 |
100 μL |
453.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
OLFM3 (olfactomedin 3), also known as NOE3, is a 478 amino acid protein that interacts with myocilin. Myocilin is an extracellular protein that plays a key role in the actomyosin system and is responsible for controlling intraocular pressure. OLFM3 is a secreted protein that contains an olfactomedin-like (OLF) domain, an approximately 260 amino acid motif commonly found in secreted glycoproteins. OLFM3 localizes to the Golgi apparatus of the cell and is highly expressed in both eye and brain tissue. Mutations in the gene that encodes OLFM3 may cause severe glaucoma, a condition in which increased intraocular pressure within the eyeball causes loss of eye sight.Specifications
| OLFM3 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q96PB7 | |
| 118427 | |
| Gln24-Glu198 (Accession Q96PB7), with N-terminal His Tag. | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Olfm3 |
| 0.5 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| B230206G02Rik; NOE3; NOELIN3; Noelin-3; olfactomedin 3; olfactomedin related ER localized protein 3; olfactomedin-3; OLFM3; OPTIMEDIN; UNQ1924/PRO4399 | |
| Olfm3 | |
| IgG | |
| Antigen affinity chromatography, Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title