Learn More
Cytiva Omics Applicator
Product Code. 30057715 Shop All Cytiva Products
Change view
Click to view available options
For Use With (Equipment):
Tissue Disaggregator
Unit Size:
Pack of 60
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
30057715 Tissue Disaggregator Pack of 60
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30057715 Supplier Cytiva Supplier No. 29509359
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Open-bore sample insertion syringe to aid insertion of tissue sample into Omics pouch.
  • For use with Omics pouch
TRUSTED_SUSTAINABILITY
Specifications
Product Type Applicator
Autoclavable Not Autoclavable
Closure Color White
For Use With (Equipment) Tissue Disaggregator
For research use only (RUO). Not for diagnostic use.