Learn More
Cytiva VIA Extractor™ tissue disaggregator - Omics Bundle
Product Code. 30057808 Shop All Cytiva Products
Change view
Click to view available options
Voltage:
24 V
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Voltage Unit Size
30057808 24 V 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is under eco-contribution tax.
Product Code. 30057808 Supplier Cytiva Supplier No. 29517120
Estimated Shipment: 15-04-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: This product is under eco-contribution tax.
Semi-automated system for consistent, rapid disaggregation of solid tissue, including tumors, into viable single cells. The VIA Extractor includes tissue disaggregator, controlled-rate freezer and the Omics clamp.
  • The VIA Extractor™ tissue disaggregator disaggregates solid tissue into viable single cells
  • For use in high-throughput omics and flow cytometry research
  • The closed system uses mild processing to optimize yield and preserve content relative to the parent sample
TRUSTED_SUSTAINABILITY
Specifications
Product Type Tissue Disaggregator
Speed ≤460 rpm
Voltage 24 V
For research use only (RUO). Not for diagnostic use.