Learn More
Diba Omnifit™ BenchMark™ Replacement Frits for Chromatography Column Assemblies
Product Code. 15895524
Change view
Click to view available options
Porosity:
5 μm
10 μm
25 μm
Diameter (Metric) Inner:
15 mm
35 mm
50 mm
Material:
PTFE
Polyethylene
Unit Size:
Pack of 20
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Porosity Diameter (Metric) Inner Material Unit Size
15895524 25 μm 15 mm Polyethylene Pack of 20
15845544 5 μm 50 mm PTFE Pack of 20
15815544 25 μm 35 mm Polyethylene Pack of 20
15855544 10 μm 50 mm PTFE Pack of 20
15875544 25 μm 50 mm Polyethylene Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 15895524 Supplier Diba Supplier No. 006FR1525PE
Estimated Shipment: 23-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Select from a variety of porosities for your media and chemistry
Replacement frits are available in PTFE or PE and in a range of pore sizes, enabling you to select the most appropriate bed supports for your application. Match the column ID to select the correct frit
Specifications
For Use With (Equipment) Chromatography Column Assemblies
Material Polyethylene
Porosity 25 μm
Size Universal
Diameter (Metric) Inner 15 mm
Diameter (English) Inner 0.59 in.
Type Replacement Chromatography Frit