Learn More
- Home
- Products
- Chromatography Columns and Cartridges
- Low to Medium Pressure LC Columns
- Diba Omnifit™ EZ SolventPlus™ Chromatography Columns with Two Fixed Endpieces
Diba Omnifit™ EZ SolventPlus™ Chromatography Columns with Two Fixed Endpieces
Product Code. 15805424
Change view
Click to view available options
Length (Metric):
50 mm
100 mm
150 mm
250 mm
330 mm
400 mm
440 mm
500 mm
Diameter (Metric) Inner:
6 mm
10 mm
15 mm
25 mm
35 mm
50 mm
Column Capacity:
0.7 mL
2.4 mL
4.1 mL
5.5 mL
7.5 mL
9.4 mL
10.3 mL
12.4 mL
12.7 mL
17.3 mL
21.2 mL
23.6 mL
29.1 mL
34.4 mL
36.9 mL
38.9 mL
53 mL
58.9 mL
65.4 mL
83.1 mL
108 mL
115.5 mL
147.3 mL
181.6 mL
211.7 mL
230.7 mL
288.6 mL
356 mL
440.7 mL
600.9 mL
741.1 mL
941.4 mL
Unit Size:
Each
32 product options available for selection
| Product Code. | Length (Metric) | Diameter (Metric) Inner | Column Capacity | unitSize |
|---|---|---|---|---|
| 15805424 | 50 mm | 6 mm | 0.7 mL | Each |
| 15835424 | 100 mm | 6 mm | 2.4 mL | Each |
| 15865424 | 150 mm | 6 mm | 4.1 mL | Each |
| 15895424 | 250 mm | 6 mm | 7.5 mL | Each |
| 15825434 | 330 mm | 6 mm | 10.3 mL | Each |
| 15855434 | 440 mm | 6 mm | 12.7 mL | Each |
| 15885434 | 100 mm | 10 mm | 5.5 mL | Each |
| 15815444 | 150 mm | 10 mm | 9.4 mL | Each |
| 15845444 | 250 mm | 10 mm | 17.3 mL | Each |
| 15875444 | 330 mm | 10 mm | 23.6 mL | Each |
| 15805454 | 400 mm | 10 mm | 29.1 mL | Each |
| 15835454 | 500 mm | 10 mm | 36.9 mL | Each |
| 15865454 | 100 mm | 15 mm | 12.4 mL | Each |
| 15895454 | 150 mm | 15 mm | 21.2 mL | Each |
| 15825464 | 250 mm | 15 mm | 38.9 mL | Each |
| 15855464 | 330 mm | 15 mm | 53 mL | Each |
| 15885464 | 400 mm | 15 mm | 65.4 mL | Each |
| 15815474 | 500 mm | 15 mm | 83.1 mL | Each |
| 15845474 | 100 mm | 25 mm | 34.4 mL | Each |
| 15875474 | 150 mm | 25 mm | 58.9 mL | Each |
| 15805484 | 250 mm | 25 mm | 108 mL | Each |
| 15835484 | 330 mm | 25 mm | 147.3 mL | Each |
| 15865484 | 400 mm | 25 mm | 181.6 mL | Each |
| 15895484 | 500 mm | 25 mm | 230.7 mL | Each |
| 15825494 | 150 mm | 35 mm | 115.5 mL | Each |
| 15855494 | 250 mm | 35 mm | 211.7 mL | Each |
| 15885494 | 330 mm | 35 mm | 288.6 mL | Each |
| 15815504 | 400 mm | 35 mm | 356 mL | Each |
| 15845504 | 250 mm | 50 mm | 440.7 mL | Each |
| 15875504 | 330 mm | 50 mm | 600.9 mL | Each |
| 15805514 | 400 mm | 50 mm | 741.1 mL | Each |
| 15835514 | 500 mm | 50 mm | 941.4 mL | Each |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 32 options available.
Description
- PEEK™ column threads offer outstanding durability
- Superior frit placement adds bed support and even distribution of sample flow
- Reusable columns let you pack your own media specific to your application
Specifications
Specifications
| Temperature (Metric) Range | 4 to 40°C |
| Diameter (Metric) Inner | 6 mm |
| Type | Liquid Chromatography Column |
| Max. Pressure | 900 psi |
| Pore Size | 10 and 30 μm |
| Column Capacity | 0.7 mL |
| Length (Metric) | 50 mm |
| Material | Borosilicate Glass, PTFE, Chemraz™ |
| Max. Temperature | 40°C |
| Includes | Two (0.5 m and 2 m L) 1/16 in. OD PTFE tubing assembly with 1/4-28 UNF ETFE fitting at one end, a frit kit (contains two each of 10 μm PE, 10 μm PTFE, and 30 μm PTFE frits) and a Click-N-Seal™ 10-32 PEEK conical port fitting. |