Learn More
- Home
- Products
- Chromatography Columns and Cartridges
- Low to Medium Pressure LC Columns
- Diba Omnifit™ EZ SolventPlus™ Chromatography Columns with Two Fixed Endpieces
Diba Omnifit™ EZ SolventPlus™ Chromatography Columns with Two Fixed Endpieces
Product Code. 15815444
Change view
Click to view available options
Length (Metric):
50 mm
100 mm
150 mm
250 mm
330 mm
400 mm
440 mm
500 mm
Diameter (Metric) Inner:
6 mm
10 mm
15 mm
25 mm
35 mm
50 mm
Column Capacity:
0.7 mL
2.4 mL
4.1 mL
5.5 mL
7.5 mL
9.4 mL
10.3 mL
12.4 mL
12.7 mL
17.3 mL
21.2 mL
23.6 mL
29.1 mL
34.4 mL
36.9 mL
38.9 mL
53 mL
58.9 mL
65.4 mL
83.1 mL
108 mL
115.5 mL
147.3 mL
181.6 mL
211.7 mL
230.7 mL
288.6 mL
356 mL
440.7 mL
600.9 mL
741.1 mL
941.4 mL
Unit Size:
Each
32 product options available for selection
| Codice prodotto. | Length (Metric) | Diameter (Metric) Inner | Column Capacity | Dimensione della confezione |
|---|---|---|---|---|
| 15815444 | 150 mm | 10 mm | 9.4 mL | Each |
| 15805424 | 50 mm | 6 mm | 0.7 mL | Each |
| 15835424 | 100 mm | 6 mm | 2.4 mL | Each |
| 15865424 | 150 mm | 6 mm | 4.1 mL | Each |
| 15895424 | 250 mm | 6 mm | 7.5 mL | Each |
| 15825434 | 330 mm | 6 mm | 10.3 mL | Each |
| 15855434 | 440 mm | 6 mm | 12.7 mL | Each |
| 15885434 | 100 mm | 10 mm | 5.5 mL | Each |
| 15845444 | 250 mm | 10 mm | 17.3 mL | Each |
| 15875444 | 330 mm | 10 mm | 23.6 mL | Each |
| 15805454 | 400 mm | 10 mm | 29.1 mL | Each |
| 15835454 | 500 mm | 10 mm | 36.9 mL | Each |
| 15865454 | 100 mm | 15 mm | 12.4 mL | Each |
| 15895454 | 150 mm | 15 mm | 21.2 mL | Each |
| 15825464 | 250 mm | 15 mm | 38.9 mL | Each |
| 15855464 | 330 mm | 15 mm | 53 mL | Each |
| 15885464 | 400 mm | 15 mm | 65.4 mL | Each |
| 15815474 | 500 mm | 15 mm | 83.1 mL | Each |
| 15845474 | 100 mm | 25 mm | 34.4 mL | Each |
| 15875474 | 150 mm | 25 mm | 58.9 mL | Each |
| 15805484 | 250 mm | 25 mm | 108 mL | Each |
| 15835484 | 330 mm | 25 mm | 147.3 mL | Each |
| 15865484 | 400 mm | 25 mm | 181.6 mL | Each |
| 15895484 | 500 mm | 25 mm | 230.7 mL | Each |
| 15825494 | 150 mm | 35 mm | 115.5 mL | Each |
| 15855494 | 250 mm | 35 mm | 211.7 mL | Each |
| 15885494 | 330 mm | 35 mm | 288.6 mL | Each |
| 15815504 | 400 mm | 35 mm | 356 mL | Each |
| 15845504 | 250 mm | 50 mm | 440.7 mL | Each |
| 15875504 | 330 mm | 50 mm | 600.9 mL | Each |
| 15805514 | 400 mm | 50 mm | 741.1 mL | Each |
| 15835514 | 500 mm | 50 mm | 941.4 mL | Each |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 32 options available.
Denna artikel kan inte returneras.
Se returpolicy
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
- PEEK™ column threads offer outstanding durability
- Superior frit placement adds bed support and even distribution of sample flow
- Reusable columns let you pack your own media specific to your application
Spezifikation
Spezifikation
| Max. Pressure | 600 psi |
| Pore Size | 10 and 30 μm |
| Column Capacity | 9.4 mL |
| Length (Metric) | 150 mm |
| Material | Borosilicate Glass, PTFE, Chemraz™ |
| Max. Temperature | 40°C |
| Includes | Two (0.5 m and 2 m L) 1/16 in. OD PTFE tubing assembly with 1/4-28 UNF ETFE fitting at one end, a frit kit (contains two each of 10 μm PE, 10 μm PTFE, and 30 μm PTFE frits) and a Click-N-Seal™ 10-32 PEEK conical port fitting. |
| Temperature (Metric) Range | 4 to 40°C |
| Type | Liquid Chromatography Column |
| Diameter (Metric) Inner | 10 mm |
Name des Produkts
Wählen Sie ein Thema
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Haben Sie Verbesserungsvorschläge?