Learn More
Ondansetron hydrochloride dihydrate, 98%
Product Code. 15678715 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15678715 1 g 1g
15688715 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15678715 Supplier Thermo Scientific Acros Supplier No. 461160010
 Restricted Item
In Stock
Add to basket
missing translation for 'validMainframeMsg'
CAS: 103639-04-9 | HCl·2H2O | 365.85 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 103639-04-9
Molecular Formula HCl·2H2O
Molecular Weight (g/mol) 365.85
InChI Key BGGIFKYQGVGSIW-UHFFFAOYSA-N
Synonym 1,2,3,9-tetrahydro-9-methyl-3-2-methyl-1h-imidazol-1-yl methyl-4h-carbazol-4-one hydrochloride; gr 38032f; ondansetron hydrochloride
PubChem CID 71317260
IUPAC Name chlorine;9-methyl-3-[(2-methylimidazol-1-yl)methyl]-2,3-dihydro-1H-carbazol-4-one;dihydrate
SMILES CC1=NC=CN1CC2CCC3=C(C2=O)C4=CC=CC=C4N3C.O.O.[Cl]
Specifications
Melting Point 178.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 365.85
Percent Purity 98%
Chemical Name or Material Ondansetron hydrochloride dihydrate
compliance-icons compliance-icons
Product Identifier
  • Ondansetron hydrochloride dihydrate, 98%
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Supplemental information
  • MIXTURE LIST-Contain: Ondansetron hydrochloride dihydrate