Learn More
Hamilton™ One Hub Assembled with Custom Length Tubing (1/8in.)
Product Code. 11533134
Change view
Click to view available options
Overall Length (English):
Custom Length
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Overall Length (English) Unit Size
11533134 Custom Length Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11533134 Supplier Hamilton™ Supplier No. 90691
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
PTFE tubing has high chemical resistance, low and high temperature capability, resistance to weathering, low friction, and electrical and thermal insulation.
Specification Sheet
  • Fitting Material-CTFE
  • Tubing Material-PTFEGauge1/8 in.
  • Length-Custom Length
  • Sales Quantity-1 each
  • Fitting Type-Female Luer
  • Number of Fittings-one
Specifications
Diameter (English) Inner 1/8 in.
Overall Length (English) Custom Length
Material PTFE