missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OPALIN Antibody (CL11144) [PerCP], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07995PCP
This item is not returnable.
View return policy
Description
OPALIN Monoclonal antibody specifically detects OPALIN in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| OPALIN | |
| Monoclonal | |
| PerCP | |
| HTMP10transmembrane protein TMP10, oligodendrocytic myelin paranodal and inner loop proteinTMEM10, TMP10, Transmembrane protein 10opalin | |
| This antibody was developed using a recombinant protein derived from Q96PE5, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL11144 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 93377 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction