missing translation for 'onlineSavingsMsg'
Learn More

OPC 14714, TRC

Product Code. 30418110
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30418110 10 mg 10mg
30413009 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30418110 Supplier Toronto Research Chemicals Supplier No. TRCO66760010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 203395-84-0
Molecular Formula C23 H28 Br N3 O2
Molecular Weight (g/mol) 458.39
Synonym 2(1H)-Quinolinone, 7-[4-[4-(2-bromophenyl)-1-piperazinyl]butoxy]-3,4-dihydro- (9CI, ACI), 7-[4-[4-(2-Bromophenyl)-1-piperazinyl]butoxy]-3,4-dihydro-2(1H)-quinolinone (ACI), 7-[4-[4-(2-bromophenyl)piperazin-1-yl]butoxy]-3,4-dihydroquinolin-2(1H)-one, 2-Monobromo Aripiprazole, 2-Monobromoaripiprazole
IUPAC Name 7-[4-[4-(2-bromophenyl)piperazin-1-yl]butoxy]-3,4-dihydro-1H-quinolin-2-one
SMILES Brc1ccccc1N2CCN(CCCCOc3ccc4CCC(=O)Nc4c3)CC2

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C23H28BrN3O2/c24-20-5-1-2-6-22(20)27-14-12-26(13-15-27)11-3-4-16-29-19-9-7-18-8-10-23(28)25-21(18)17-19/h1-2,5-7,9,17H,3-4,8,10-16H2,(H,25,28)
Formula Weight 457.1365
Chemical Name or Material OPC-14714
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.