Learn More
Berkshire™ Open-Cell Foam Lab-Tips™ Swabs
Product Code. 17833232 Shop All Berkshire Products
Change view
Click to view available options
Length (Metric):
71 mm
76 mm
127 mm
162 mm
Width (Metric) Tip:
3 mm
3.7 mm
3.8 mm
4 mm
5 mm
13 mm
Unit Size:
Each
Pack of 1000
Pack of 20
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Width (Metric) Tip unitSize
17833232 76 mm 5 mm Each
17897290 76 mm 5 mm Pack of 1000
17863232 71 mm 4 mm Each
17825510 71 mm 3 mm Pack of 20
17873232 71 mm 3 mm Each
12876774 71 mm 3.7 mm Pack of 20
17883232 71 mm 3.7 mm Each
17853232 162 mm 3.8 mm Each
12897153 127 mm 13 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 missing translation for 'options'
Product Code. 17833232 missing translation for 'mfr' Berkshire™ missing translation for 'supplierNo' LTA03163.10
  • ONLINE EXCLUSIVE PRICE
    796.95€
    763.92€ / Pack of 1
    Save 33.03€
    4% Off
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
Add to basket
General-purpose swab for cleaning small and confined areas
  • Polyurethane foam over rigid core
  • 100 ppi open-cell foam
  • Thermally bonded head
  • Low particles, fibers, ions and extractables, no contaminating adhesives
  • Good chemical resistance
  • Good sorbency and excellent solvent hold
  • Laundered and packaged in cleanroom
Specifications
Certifications/Compliance ISO Class 4
Diameter (English) Handle 0.18 in.
Length (English) 3 in.
For Use With (Application) For cleaning small and confined areas, good chemical resistance, sorbency and excellent solvent hold
Product Type Swab
Diameter (Metric) Handle 4.5 mm
Length (Metric) 76 mm
Length (English) Handle 2.8 in.
Length (Metric) Handle 71 mm
Material Polyurethane Foam
Shape Sock Tip
Thickness (Metric) 3 mm
Length (English) Tip 0.5 in.
Handle Type Rigid
Length (Metric) Tip 13 mm
Width (Metric) Tip 5 mm
Width (English) Tip 0.19 in.
Handle Material Polypropylene
Tip Material Polyurethane Foam