Learn More
OPLAH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21127-1-AP-20UL
This item is not returnable.
View return policy
Description
OPLAH catalyzes the cleavage of 5-oxo-L-proline to form L-glutamate coupled to the hydrolysis of ATP to ADP and inorganic phosphate. ATP + 5-oxo-L-proline + 2 H2O = ADP + phosphate + L-glutamate.Specifications
| OPLAH | |
| Polyclonal | |
| Unconjugated | |
| OPLAH | |
| 5 Opase, 5 oxo L prolinase, 5 oxoprolinase, OPLA, OPLAH, Pyroglutamase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 116684, 26873, 75475 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O14841, P97608, Q8K010 | |
| Oplah | |
| OPLAH Fusion Protein Ag15523 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |