Learn More
Hellma™ Optical Glass Micro Cells
Product Code. 10673711
Change view
Click to view available options
Capacity (Metric):
7 mL
17.5 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10673711 17.5 mL Each
10343841 7 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10673711 Supplier Hellma™ Supplier No. 60305010
In Stock
Add to basket
Edge
Add to basket
Specifications
Type Micro Cell
Capacity (Metric) 17.5 mL
Material Optical Glass
Closure Material PTFE
Width (Metric) Interior 9.5mm
Quantity 1Each
Path Length 50 mm
Wavelength Range 360 to 2500 nm
Height (Metric) 45 mm
Width (Metric) 12.5 mm