Learn More
Olympus™ Lens Cleaning Optical Paper
Product Code. 12529225
Change view
Click to view available options
Length (Metric):
70 mm
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
12529225 70 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12529225 Supplier Olympus™ Supplier No. K10017026
  • ONLINE EXCLUSIVE PRICE
    32.70€
    14.23€ / Pack of 100
    Save 18.47€
    56% Off
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Suitable lens cleaning tissues for cleaning all kinds of glass surfaces including multi-coated precision lenses. Olympus™ Lens Cleaning Optical Paper keep your optics clean and your image data crisp and accurate.
  • High quality lens paper
  • Preserves the quality of your microscope optics
  • 100 sheets/pack
Specifications
Type Optical Paper
Dimensions (L x W x H) 70 x 120 mm
For Use With (Application) For lens cleaning
Length (Metric) 70 mm
Material Optical Paper
Quantity 100 Pak.
Width (Metric) 120 mm
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.