Learn More
SHP Steriltechnik AG Option V Laboklav 25, Vacuum
Description
- Permanent or fractional pre-vacuum for optimal air removal
- Safe penetration of steam into sterilisation items that are difficult to vent
- Reproducible sterilisation results in accordance with DIN 58951-2
- Drying vacuum for efficient post-drying of packaged items
- Condensate is reliably removed from the packaging
- Avoids the need for additional transfer to a drying cabinet
- Suitable for solid, moist or packaged items
Specifications
Specifications
| For Use With (Equipment) | Autoclaves, Laboklav 25, Laboklav ECO |
| Model | Option V Laboklav 25 and ECO |
| Product Type | Autoclave Vacuum |