missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OPTIZYME™ XmaJI (AvrII), Fisher BioReagents™
Brand: Fisher Bioreagents BP8082-1
Additional Details : Weight : 0.01000kg
Description
5'...C^C T A G G...3'
3'...G G A T C^C...5'
Supplied with: 10X OPTIZYME 4
Conditions for 100% Activity:
- 1X OPTIZYME Buffer 4: 33mM Tris-acetate (pH 7.9 at 37°C), 10mM Mg-acetate, 66mM K-acetate and 0.1mg/mL BSA
- Incubate at 37°C
- Buffer 1: 20 - 50%
- Buffer 2: 50 -100%
- Buffer 3: 50 - 100%
- Buffer 4: 100%
- Buffer 5: 50 - 100%
Storage Buffer:
- 10mM Tris-HCl (pH 7.4 at 25°C), 100mM KCl, 1mM DTT, 1mM EDTA, 0.2mg/mL BSA and 50% (v/v) glycerol
Specifications
10 U/μL | |
37°C | |
>95% of DNA fragments can be ligated and re-cut after 50 fold over-digestion with XmaJI | |
200 U | |
XmaJI (AvrII) |
10X OPTIZYME™ Buffer 4 | |
7.4 | |
10mM Tris HCl (pH 7.4 at 25°C), 100mM KCl, 1mM DTT, 1mM EDTA, 0.2mg/mL BSA, 50% Glycerol | |
C.CTAGG | |
Liquid |