missing translation for 'onlineSavingsMsg'
Learn More

Oregon Pesticide Mixture 6 600 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30321695
Change view
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30321695 5 x 1 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30321695 Supplier Dr. Ehrenstorfer Supplier No. DREGS09000236AL

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acephate∣Chlorpyrifos∣Diazinon∣Dichlorvos∣Dimethoate∣Ethoprophos∣Malathion∣Naled∣Parathion-methyl∣Phosmet
CAS 30560-19-1, 2921-88-2, 333-41-5, 62-73-7, 60-51-5, 13194-48-4
Analyte or Component Names Acephate∣Chlorpyrifos∣Diazinon∣Dichlorvos∣Dimethoate∣Ethoprophos∣Malathion∣Naled∣Parathion-methyl∣Phosmet
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Acetonitrile
Quantity 5 x 1 mL
Concentration or Composition (by Analyte or Components) Acephate 600 μg/mL∣Chlorpyrifos 600 μg/mL∣Diazinon 600 μg/mL∣Dichlorvos 600 μg/mL∣Dimethoate 600 μg/mL∣Ethoprophos 600 μg/mL∣Malathion 600 μg/mL∣Naled 600 μg/mL∣Parathion-methyl 600 μg/mL∣Phosmet 600 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.