missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Orexin Receptor 1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA577566
This item is not returnable.
View return policy
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 μL of deionized water.
The protein encoded by this gene is a G-protein coupled receptor involved in the regulation of feeding behavior. The encoded protein selectively binds the hypothalamic neuropeptide orexin A. A related gene (HCRTR2) encodes a G-protein coupled receptor that binds orexin A and orexin B.
Specifications
| Orexin Receptor 1 | |
| Polyclonal | |
| Unconjugated | |
| HCRTR1 | |
| OX1R, HCRTR1, Hypocretin receptor 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 230777, 25593, 3061 | |
| -20°C | |
| Lyophilized |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
| 0.8 mg/mL | |
| PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
| O43613, P56718, P58307 | |
| Hcrtr1 | |
| Peptide (C)RNWKRPSEQLEAQH, corresponding to amino acid residues 256-269 of rat OX1 R | |
| 50 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction