missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Orexin receptor 1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18370-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a G-protein coupled receptor involved in the regulation of feeding behavior. The encoded protein selectively binds the hypothalamic neuropeptide orexin A. A related gene (HCRTR2) encodes a G-protein coupled receptor that binds orexin A and orexin B.Specifications
| Orexin receptor 1 | |
| Polyclonal | |
| Unconjugated | |
| HCRTR1 | |
| HCRTR1, hypocretin (orexin) receptor 1, Hypocretin receptor type 1, Orexin receptor 1, Orexin receptor type 1, Ox 1 R, Ox1 R, OX1R | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 230777, 25593, 3061 | |
| -20°C | |
| Liquid |
| Western Blot, Flow Cytometry | |
| 0.22 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O43613, P56718, P58307 | |
| Hcrtr1 | |
| Orexin receptor 1 Fusion Protein Ag12269 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu