missing translation for 'onlineSavingsMsg'
Learn More

ORF73/HHV8 Antibody (LN53), PE, Novus Biologicals™

Rat Monoclonal Antibody

Brand:  Novus Biologicals NBP3-08217PE

Product Code. 18600899

  • 748.00€ / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

ORF73/HHV8 Monoclonal antibody specifically detects ORF73/HHV8 in Nonspecies specific samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ORF73/HHV8
Monoclonal
PE
HHV 8, Human Herpes Virus 8, Kaposi's sarcoma associated herpes virus, KSHV
Recombinant protein corresponding to the latent nuclear antigen 1 molecule of ORF73/HHV8
0.1 mL
Adaptive Immunity, Immunology, Virology Bacteria and Parasites
935114
Store at 4°C in the dark.
IgG2c κ
ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
LN53
PBS
Rat
Protein A or G purified
RUO
Primary
Non-species specific
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.