missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ORM1 Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA5149098
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
ORM1 is an acute phase plasma protein synthesized by the liver. The protein is believed to regulate the interaction between blood cells and endothelial cells, and together with haptoglobin and C reactive protein, also regulates the extravasation of the cells during infection and inflammation. ORM1 is a major acute phase reactant, both in human and in rat. In human plasma, ORM1 (44 kDa) is found at levels of 0.5-1.4 mg/mL. Its concentration in blood plasma is elevated during acute inflammation, such as trauma, malignancies, myocardial infarction, rheumatoid arthritis, bacterial infections, after major surgery, in cases of chronic pain etc, and can be used for the diagnosis of inflammatory conditions.
Specifications
| ORM1 | |
| Polyclonal | |
| Unconjugated | |
| ORM1 | |
| AGP; AGP 1; Agp1; Agp-1; Agp-2; AGP-A; Agpa1; alpha-1 acid glycoprotein; alpha-1-acid glycoprotein; Alpha-1-acid glycoprotein 1; alpha-1-acid glycoprotein-2; bM340H1.2 (orosomucoid 1); epididymis secretory sperm binding protein Li 153w; HEL-S-153w; OMD; OMD 1; Orm; Orm1; Orm-1; Orosomucoid; orosomucoid 1; orosomucoid-1 | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 18405 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q60590 | |
| ORM1 | |
| Mouse plasma-derived alpha 1-Acid Glycoprotein. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction