missing translation for 'onlineSavingsMsg'
Learn More

Orthanilic acid, 95%

Product Code. 15484387
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15484387 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15484387 Supplier Thermo Scientific Alfa Aesar Supplier No. H31763.22
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 88-21-1 | C6H7NO3S | 173.186 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 88-21-1
Melting Point >300°C
Odor Odorless
Molecular Formula C6H7NO3S
MDL Number MFCD00007705
Quantity 100 g
Synonym orthanilic acid, aniline-2-sulfonic acid, o-aminobenzenesulfonic acid, o-sulfanilic acid, aniline-o-sulfonic acid, 2-sulfanilic acid, aniline-o-sulphonic acid, 1-aminobenzene-2-sulfonic acid, 2-aminobenzenesulphonic acid, benzenesulfonic acid, 2-amino
InChI Key ZMCHBSMFKQYNKA-UHFFFAOYSA-N
SMILES C1=CC=C(C(=C1)N)S(=O)(=O)O
IUPAC Name 2-aminobenzenesulfonic acid
Molecular Weight (g/mol) 173.186
PubChem CID 6926
ChEBI CHEBI:1015
Formula Weight 173.19
Percent Purity 97%
Chemical Name or Material Orthanilic acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Orthanilic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Orthanilic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.