missing translation for 'onlineSavingsMsg'
Learn More

ortho-Topolin Riboside, TRC

Product Code. 30457315
Change view
Click to view available options
Quantity:
10 mg
50 mg
250 mg
Unit Size:
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30457315 50 mg 50mg
30454113 250 mg -
30457180 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30457315 Supplier Toronto Research Chemicals Supplier No. TRCT54028550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50868-58-1
Molecular Formula C17H19N5O5
Molecular Weight (g/mol) 373.36
Synonym 6-(2-Hydroxybenzylamino)-9-ß-D-ribofuranosylpurine, ortho-Topolin-9-riboside, N6-(o-Hydroxybenzyl)adenosine, Cytokinin R, 6-(o-Hydroxybenzylamino)-9-beta-D-ribofuranosylpurine, 6-(2-Hydroxybenzyl)aminopurine Riboside, 6-(2-Hydroxybenzyl)adenosine, (o-Hydroxybenzyl)adenine Riboside, N-[(2-Hydroxyphenyl)methyl]adenosine
IUPAC Name (2R,3S,4R,5R)-2-(hydroxymethyl)-5-[6-[(2-hydroxyphenyl)methylamino]purin-9-yl]oxolane-3,4-diol
SMILES OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)n2cnc3c(NCc4ccccc4O)ncnc23

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C17H19N5O5/c23-6-11-13(25)14(26)17(27-11)22-8-21-12-15(19-7-20-16(12)22)18-5-9-3-1-2-4-10(9)24/h1-4,7-8,11,13-14,17,23-26H,5-6H2,(H,18,19,20)/t11-,13-,14-,17-/m1/s1
Formula Weight 373.1386
Percent Purity >95%
Chemical Name or Material ortho-Topolin Riboside
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.