Learn More
Osmometers
Product Code. 11900557
Change view
Click to view available options
Display Type:
LCD Graphic
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Display Type Unit Size
11900557 LCD Graphic Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11900557 Supplier Loeser Supplier No. TYP15
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Specifications
Display Type LCD Graphic
Frequency 50 Hz
Voltage 230 V
Cooling Agent Peltier Effect
Depth (Metric) 216 mm
Quantity 1 Ea.
Product Type Osmometer
Model Type 15
For Use With (Application) Food: milk water content. Hospital: control injections. Transfusion Centre: control of physicochemical properties chemical derivatives of blood and analyzed for residual glycerol after thawing. Cell culture, biochemistry, marine biology, plant physiology
Linearity Raised By Higher Concentrations
Height (Metric) 278 mm
Measurement Time 1.5 min.
Power Requirements 230 VAC (100/115 VAC on request) approx. 45 VA, 50 Hz, 20/45 w
Range 0 to 2500 mosm/kg
Resolution 1 mOsm/kg
Sample Size 50 to 100 μL
Wattage 20/45 w
Weight (Metric) 6.3 kg
Width (Metric) 180 mm
Electrical Requirements 230 VAC (100/115 VAC on request) approx. 45 VA, 50 Hz, 20/45 w