missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OSTF1 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-50036PCP
This item is not returnable.
View return policy
Description
OSTF1 Polyclonal antibody specifically detects OSTF1 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| OSTF1 | |
| Polyclonal | |
| Western Blot, Immunoprecipitation | |
| FLJ20559, OSFSH3P2bA235O14.1, osteoclast stimulating factor 1, osteoclast-stimulating factor 1 | |
| The immunogen for this product maps to a region between residue 50 and 100 of human Osteoclast Stimulating Factor 1 using the numbering given in entry NP_036515.4 (GeneID 26578). | |
| 0.1 mL | |
| Signal Transduction | |
| 26578 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| PerCP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction