missing translation for 'onlineSavingsMsg'
Learn More

Osutidine Hydrochloride Salt, TRC

Product Code. 30421816
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30421816 50 mg 50mg
30409929 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30421816 Supplier Toronto Research Chemicals Supplier No. TRCO70391050MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Osutidine Hydrochloride Salt
Molecular Formula C19H28N4O5S2 . HCl
Quantity 50 mg
InChI Formula InChI=1S/C19H28N4O5S2.ClH/c1-20-11-16-7-8-17(28-16)13-29-10-9-21-19(23-30(2,26)27)22-12-18(25)14-3-5-15(24)6-4-14;/h3-8,18,20,24-25H,9-13H2,1-2H3,(H2,21,22,23);1H
SMILES CNCC1=CC=C(CSCCN/C(NCC(C2=CC=C(O)C=C2)O)=N/S(C)(=O)=O)O1.Cl
IUPAC Name (E)-N-(((2-hydroxy-2-(4-hydroxyphenyl)ethyl)amino)((2-(((5-((methylamino)methyl)furan-2-yl)methyl)thio)ethyl)amino)methylene)methanesulfonamide hydrochloride
Molecular Weight (g/mol) 493.04
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.