Learn More
OTC Monoclonal Antibody (OTI8H9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA802457S
Description
OTC Monoclonal antibody specifically detects OTC in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| OTC | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P00480 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 5009 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI8H9 | |
| Unconjugated | |
| OTC | |
| AI265390; OCTD; ornithine carbamoyltransferase; ornithine carbamoyltransferase, mitochondrial; Ornithine transcarbamylase; Otc; OTCase; Sf; sparse fur; spf | |
| Human recombinant protein fragment corresponding to amino acids 33-354 of human OTC produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.