Learn More
OTP Antibody (CL11225) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Marque:  Novus Biologicals NBP3-15136JF525
Code produit. 30287947
  • 737.00€ / 0.10mL
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
OTP Monoclonal antibody specifically detects OTP in Human,Rat samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Spécification
Spécification
OTP
Monoclonal
Janelia Fluor 525
homeobox protein orthopedia, MGC3161, orthopedia homeobox, orthopedia homolog, orthopedia homolog (Drosophila)
This antibody was developed using a recombinant protein derived from Q5XKR4, with the exact immunogen sequence remaining proprietary.
0.1 mL
Neuroscience
23440
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL11225
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human, Rat