missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OTUD5 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76943-0.025mg
This item is not returnable.
View return policy
Description
OTUD5 Polyclonal specifically detects OTUD5 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| OTUD5 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Deubiquitinating enzyme A, DKFZp761A052, DUBAdeubiquinating enzyme A, EC 3.4.19.12, MGC104871, OTU domain containing 5, OTU domain-containing protein 5 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 55593 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL | |
| EAW50723 | |
| OTUD5 | |
| Antibody was raised against a 13 amino acid synthetic peptide near the carboxy terminus of the human OTUD5. The immunogen is located within amino acids 400 - 450 of OTUD5. Amino Acid Squence: VRGPSQPKASAT | |
| 0.025 mg | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction