Learn More
VistaLab™ Technologies Ovation™ Pipet Accessories: Reconditioning Nozzle Kit
For Ovation E8 & E12 Multichannel Pipets
Brand: VistaLab™ Technologies 9060-3011
Additional Details : Weight : 0.10000kg
Beschreibung
- Contains 2 nozzles, 24 nozzle seals, 24 filters & nozzle removal tool
Spezifikation
Large Nozzle Reconditioning Kit | |
25 to 850 μL, 25 to 1250 μL | |
2 Nozzles, 24 seals, 24 filters and tool | |
2 in. | |
5 in. |
Non-sterile | |
25 to 850 μL, 25 to 1250 μL Ovation™ BioNatural™ Multichannel Pipette | |
1 Ea. | |
2 in. |