Learn More
Fisherbrand™ A2 and A5 Large Gel Electrophoresis System Standard Comb
Product Code. 12413657
Change view
Click to view available options
Type:
Rapid Load Comb
Standard Comb
Teeth Count:
18
20
Thickness (Metric) Teeth:
1 mm
1.5 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Teeth Count Thickness (Metric) Teeth Unit Size
12413657 Standard Comb 20 1 mm Each
12413667 Rapid Load Comb 18 1.5 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12413657 Supplier Fisherbrand™ Supplier No. A220C
  • ONLINE EXCLUSIVE PRICE
    62.95€
    60.40€ / Pack of 1
    Save 2.55€
    4% Off
Estimated Shipment: 21-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Eliminate comb assembly with this assortment of heavy-duty, one-piece combs, for use with Fisherbrand™ A2 and A5 Large Gel Electrophoresis Systems.
Eliminate comb assembly with an assortment of heavy-duty, one-piece Fisherbrand™ A2 and A5 Large Gel Electrophoresis Systems Combs, for use with Owl A2 and A5 Large Gel Electrophoresis Systems.
Heavy-duty, one-piece combs eliminate comb assembly.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Fisherbrand™ A2 and A5 Large Gel Electrophoresis Systems
Type Standard Comb
Teeth Count 20
Thickness (Metric) Teeth 1 mm