Learn More
Thermo Scientific™ Owl™ C2-S Micro Electrophoresis System Combs
Description
Eliminate comb assembly with heavy-duty, one-piece Thermo Scientific™ Owl™ C2-S Micro Electrophoresis System Combs. These standard seven-tooth combs are available in two thicknesses (1.0 and 1.5mm) and are for use with the Owl™ C2-S Micro Electrophoresis System.
Heavy-duty, one-piece combs eliminate comb assembly.
Specifications
Specifications
| Description | Standard comb |
| For Use With (Equipment) | Owl™ C2-S Micro Electrophoresis System |
| Product Line | Owl |
| Type | Comb |
| Teeth Count | 7 |
| Thickness (Metric) Teeth | 1.5 mm |