missing translation for 'onlineSavingsMsg'
Learn More

Oxaloacetic Acid, TRC

Product Code. 30137185
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137185 5 g 5g
30136060 10 g 10g
30006657 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137185 Supplier Toronto Research Chemicals Supplier No. TRCO8450305G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 328-42-7
Molecular Formula C4 H4 O5
Molecular Weight (g/mol) 132.07
Synonym alpha-Ketosuccinic Acid, Butanedioic acid, 2-oxo- (ACI), 2-Oxobutanedioic acid (ACI), Butanedioic acid, oxo- (9CI), Oxalacetic acid (8CI), 2-Ketosuccinic acid, 2-Oxosuccinic acid, Ketosuccinic acid, NSC 284205, NSC 77688, OAA, Oxaloacetic acid, Oxaloethanoic acid, Oxosuccinic acid, α-Ketosuccinic acid
IUPAC Name 2-oxobutanedioic acid
SMILES OC(=O)CC(=O)C(=O)O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C4H4O5/c5-2(4(8)9)1-3(6)7/h1H2,(H,6,7)(H,8,9)
Formula Weight 132.0059
Percent Purity >95%
Chemical Name or Material Oxaloacetic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.