missing translation for 'onlineSavingsMsg'
Learn More

Oxazole-5-carboxylic acid, 98+%

Product Code. 15423887
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
15423887 5 g 5g
15413887 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 15423887

Brand: Thermo Scientific Alfa Aesar H30179.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 118994-90-4 | C4H3NO3 | 113.072 g/mol

Decarboxylative cross-coupling of thiazole and oxazole-5-carboxylic acids with aryl halides is reported.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Decarboxylative cross-coupling of thiazole and oxazole-5-carboxylic acids with aryl halides is reported.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118994-90-4
Molecular Formula C4H3NO3
Molecular Weight (g/mol) 113.072
MDL Number MFCD04114931
InChI Key QCGMEWVZBGQOFN-UHFFFAOYSA-N
Synonym oxazole-5-carboxylic acid, 5-oxazolecarboxylic acid, 5-oxazolecarboxylicacid, 5-carboxy-1,3-oxazole, oxazole-5-carboxylicacid, 5-carboxyoxazole, acmc-209a0v, ksc173g8b, 1,3-oxazole-5-carboxylic acid
PubChem CID 16340557
IUPAC Name 1,3-oxazole-5-carboxylic acid
SMILES C1=C(OC=N1)C(=O)O

Specifications

Melting Point ∽245°C (decomposition)
Quantity 5 g
Solubility Information Soluble in water.
Formula Weight 113.07
Percent Purity ≥98%
Chemical Name or Material Oxazole-5-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.