Learn More
Oxazole-5-carboxylic acid, 98+%
Product Code. p-7030539
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15413887 1 g 1g
15423887 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15413887 Supplier Thermo Scientific Alfa Aesar Supplier No. H30179.03
Add to basket
CAS: 118994-90-4 | C4H3NO3 | 113.072 g/mol
Decarboxylative cross-coupling of thiazole and oxazole-5-carboxylic acids with aryl halides is reported.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Decarboxylative cross-coupling of thiazole and oxazole-5-carboxylic acids with aryl halides is reported.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 118994-90-4
Molecular Formula C4H3NO3
Molecular Weight (g/mol) 113.072
MDL Number MFCD04114931
InChI Key QCGMEWVZBGQOFN-UHFFFAOYSA-N
Synonym oxazole-5-carboxylic acid, 5-oxazolecarboxylic acid, 5-oxazolecarboxylicacid, 5-carboxy-1,3-oxazole, oxazole-5-carboxylicacid, 5-carboxyoxazole, acmc-209a0v, ksc173g8b, 1,3-oxazole-5-carboxylic acid
PubChem CID 16340557
IUPAC Name 1,3-oxazole-5-carboxylic acid
SMILES C1=C(OC=N1)C(=O)O
Specifications
Melting Point ∽245°C (decomposition)
Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 113.07
Percent Purity ≥98%
Chemical Name or Material Oxazole-5-carboxylic acid