missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OXCT1 Rabbit anti-Human, Mouse, Rat, Clone: 4K12, Proteintech
Rabbit Recombinant Antibody
149.00€ - 439.00€
Specifications
| Antigen | OXCT1 |
|---|---|
| Clone | 4K12 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Immunofluorescence, ELISA |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17669816
|
Proteintech
81011-1-RR-20UL |
20 μL |
149.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17659816
|
Proteintech
81011-1-RR-100UL |
100 μL |
439.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
OXCT1 antibody apecifically detetects OXCT1 antigen in Human, Mouse, Rat Samples. It is validated for Western Blot, Immunofluorescence, ELISA.Specifications
| OXCT1 | |
| 1000 μg/mL | |
| Unconjugated | |
| Rabbit | |
| PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
| 5019 | |
| IgG | |
| Protein A purification |
| 4K12 | |
| Western Blot, Immunofluorescence, ELISA | |
| Liquid | |
| Human, Mouse, Rat | |
| BC009001 | |
| OXCT1 | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title