missing translation for 'onlineSavingsMsg'
Learn More

Oxindole (2-Indolone), TRC

Product Code. 30137008
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
Unit Size:
25g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137008 25 g 25g
30134461 50 g 50g
17901865 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137008 Supplier Toronto Research Chemicals Supplier No. TRCI62730025G
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-48-3
Molecular Formula C8 H7 N O
Molecular Weight (g/mol) 133.15
Synonym 1,3-Dihydro-2H-indol-2-one, Diclofenac Sodium Imp. E (EP), 2-Indolinone (7CI,8CI), Oxindole (6CI), 1,3-Dihydroindol-2-one, 2-Indolone, 2-Oxindole, 2-Oxo-2,3-dihydroindole, 2-Oxoindole, 2-Oxoindoline, Indol-2(3H)-one, Indoline-2-one, NSC 274863, Oxindol
IUPAC Name 1,3-dihydroindol-2-one
SMILES O=C1Cc2ccccc2N1

Specifications

Quantity 25 g
InChI Formula InChI=1S/C8H7NO/c10-8-5-6-3-1-2-4-7(6)9-8/h1-4H,5H2,(H,9,10)
Formula Weight 133.0528
Chemical Name or Material 2-Indolinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.