Learn More
Thermo Scientific™ Oxoid™ Broad Spectrum Mixture Beta Lactamase
Product Code. 13275379
Change view
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13275379 10/Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13275379 Supplier Thermo Scientific™ Supplier No. SR0113E
Mixture of beta-lactamase enzymes from Bacillus cereus for the inactivation of beta-lactam antibiotic preparations present in specimens.
Oxoid Beta Lactamase, Broad Spectrum Mixture is a mixture of beta-lactamase enzymes from Bacillus cereus for the inactivation of beta-lactam antibiotic preparations present in specimens.
TRUSTED_SUSTAINABILITY
Specifications
Description Beta Lactamase, Broad Spectrum Mixture
Form Freeze-dried
Product Type Beta Lactamase Mixture
Quantity 10/Pk.