Learn More
Thermo Scientific™ Oxoid™ Diagnostic Discs
Click to view available options
Quantity:
50 Discs
Unit Size:
Pack of 50
Description
- Differentiation of Lancefield Group A Streptococci from other beta-haemolytic Streptococci Size: 50 discs
Order Info
Supplied in cartridges of 50 discs each
Specifications
Specifications
| Antimicrobial Agents | Bacitracin |
| Quantity | 50 Discs |