missing translation for 'onlineSavingsMsg'
Learn More

Oxyclozanide, 98%, Thermo Scientific Chemicals

Product Code. 15618775 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15618775 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.
Product Code. 15618775 Supplier Thermo Scientific Acros Supplier No. 460881000
 Restricted Item
Estimated Shipment: 06-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2277-92-1
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Molecular Formula C13H6Cl5NO3
Linear Formula Cl3C6H(OH)C(=O)NHC6H2(OH)Cl2
MDL Number MFCD00864507
Quantity 100 mg
Synonym oxyclozanide, oxiclozanidum, zanil, oxyclozanid, zanilox, diplin, 2,3,5-trichloro-n-3,5-dichloro-2-hydroxyphenyl-6-hydroxybenzamide, oxyclozanide inn:ban, oxyclozanidum inn-latin, oxiclozanida inn-spanish
InChI Key JYWIYHUXVMAGLG-UHFFFAOYSA-N
SMILES OC1=C(C(=O)NC2=CC(Cl)=CC(Cl)=C2O)C(Cl)=C(Cl)C=C1Cl
IUPAC Name 2,3,5-trichloro-N-(3,5-dichloro-2-hydroxyphenyl)-6-hydroxybenzamide
Molecular Weight (g/mol) 401.45
PubChem CID 16779
Formula Weight 401.46
Percent Purity 98%
Chemical Name or Material Oxyclozanide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.