missing translation for 'onlineSavingsMsg'
Learn More

Oxytetracycline hydrochloride 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30318194
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30318194 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30318194 Supplier Dr. Ehrenstorfer Supplier No. DREA15820000AL100
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Oxytetracycline Hydrochloride
CAS 2058-46-0
Analyte or Component Names Oxytetracycline Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C22 H24 N2 O9 . Cl H
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Oxytetracycline Hydrochloride, (4S,4aR,5S,5aR,6S,12aS)-4-(Dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,5,6,10,12,12a-hexahydroxy-6-methyl-1,11-dioxo-2-naphthacenecarboxamide monohydrochloride, 4-(Dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,5,6,10,12,12a-hexahydroxy-6-methyl-1,11-dioxo-2-naphthacenecarboxamide monohydrochloride, [4S-(4alpha,4aalpha,5alpha,5aalpha,6beta,12aalpha)]-4-(Dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,5,6,10,12,12a-hexahydroxy-6-methyl-1,11-dioxo-2-naphthacenecarboxamide monohydrochloride, 1-Dimethylamino-1,4,6,11,12,13,14,18-octahydro-2,5,7,11,12,14-hexahydroxy-4,6-dioxo-11-methylnaphthacene-3-carbonamide hydrochloride, 5-Hydroxytetracycline hydrochloride, 5-Hydroxytetracycline monohydrochloride, Alamycin, Aquacycline, Biocyl, Duphacycline, Engemycin, Gynamousse, Liquamycin Injectable, Maxicyklin, Mepatar, Occrycetin, Oxlopar, Oxy WS, Oxy-Dumocyclin, Oxybiocycline, Oxybiotic, Oxycyclin, Oxyject, Oxylag, Oxypan, Oxytet, Oxytetracid, Oxytetracycline hydrochloride, Oxytetrin, TM 5, Terraject, Terramycin hydrochloride, Tetra-Tabilinen, Tetramel, Tetran hydrochloride, Toxinal, Doxycycline Hyclate Impurity E as Hydrochloride, Doxycycline Impurity E as Hydrochloride
InChI Formula InChI=1S/C22H24N2O9.ClH/c1-21(32)7-5-4-6-8(25)9(7)15(26)10-12(21)17(28)13-14(24(2)3)16(27)11(20(23)31)19(30)22(13,33)18(10)29;/h4-6,12-14,17,25,27-29,32-33H,1-3H3,(H2,23,31);1H/t12-,13-,14+,17+,21-,22+;/m1./s1
SMILES Cl.CN(C)[C@H]1[C@@H]2[C@@H](O)[C@H]3C(=C(O)[C@]2(O)C(=O)C(=C1O)C(=O)N)C(=O)c4c(O)cccc4[C@@]3(C)O
IUPAC Name (4S,4aR,5S,5aR,6S,12aS)-4-(dimethylamino)-3,5,6,10,12,12a-hexahydroxy-6-methyl-1,11-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 496.89
Quantity 1 mL
Formula Weight 496.1249 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.