missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Chloroacetophenone, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | p-Chloroacetophenone |
| CAS | 99-91-2 |
| Molecular Formula | C8H7ClO |
| Quantity | 10 g |
| Synonym | 1-(4-Chlorophenyl)-1-ethanone, 1-(4-Chlorophenyl)ethanone, 1-(p-Chlorophenyl)ethanone, 1-Acetyl-4-chlorobenzene, 1-Chloro-4-acetylbenzene, 4-Acetyl-1-chlorobenzene, 4-Acetylchlorobenzene, 4-Acetylphenyl chloride, 4-Chloro-1-acetylbenzene, 4-Chlorobenzene Methyl Ketone, 4-Chlorohypnone, 4-Chlorophenyl Methyl Ketone, 4'-Chloroacetophenone, Methyl 4-Chlorophenyl Ketone, Methyl p-Chlorophenyl Ketone, NSC 6115, p-Acetylchlorobenzene, p-Chloroacetophenone, p-Chlorohypnone, p-Chlorophenyl Methyl Ketone, 1-(4-chlorophenyl)-ethanone |
| InChI Formula | InChI=1S/C8H7ClO/c1-6(10)7-2-4-8(9)5-3-7/h2-5H,1H3 |
| SMILES | CC(=O)c1ccc(Cl)cc1 |
| IUPAC Name | 1-(4-chlorophenyl)ethanone |
| Molecular Weight (g/mol) | 154.59 |
| Formula Weight | 154.0185 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction